Ricinolja USP
Leverantör: Spectrum Chemical
Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
CA208-200LTBL CA208-20LTMT CA208-4LTGL CA208-500MLGL
SPCMCA208-200LTBLEA 61075 SEK
SPCMCA208-200LTBL SPCMCA208-20LTMT SPCMCA208-4LTGL SPCMCA208-500MLGL
Ricinolja USP
Ricinolja
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: –10 °C
Densitet: 0,95 g/cm³ (20 °C)
Flampunkt: >113 °C (closed cup)
MDL Number: MFCD00130746
CAS nummer: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908