1-Methoxy-2-methyl-2-propanol ≥98%
Leverantör: Thermo Scientific
Varning
L20351.14 L20351.22
L20351.14EA 5025 SEK
L20351.14 L20351.22
1-Methoxy-2-methyl-2-propanol ≥98%
1-Methoxy-2-methyl-2-propanol
Purity: 98+% CAS No.: 3587-64-2 Molecular Formula: C5H12O2 Formula Weight: 104.15 Flash Point: 27°(80°F) Refractive Index: 1.4044 Boiling Point: 115-116° UN No.: UN1987 MDL No.: MFCD03701584
Formel: C₅H₁₂O₂
MW: 104,15 g/mol
Kokpunkt: 115…116 °C
Densitet: 0,892
Flampunkt: 27 °C (80 °F)
Storage Temperature: Omgivande
MDL Number: MFCD03701584
CAS nummer: 3587-64-2
ADR: 3,III