4-Cyano-2-fluorobenzoic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
L19722.06 L19722.14
L19722.06EA 2150 SEK
L19722.06 L19722.14
4-Cyano-2-fluorobenzoic acid 98%
4-Cyano-2-fluorobenzoic acid
Purity: 98% CAS No.: 164149-28-4 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 214-218° Beilstein Reference: 8051929 MDL No.: MFCD03094454
Formel: C₈H₄FNO₂
MW: 165.12 g/mol
Smältpunkt: 214…218 °C
Storage Temperature: Omgivande
MDL Number: MFCD03094454
CAS nummer: 164149-28-4