Copper(I) trifluoromethanesulphonate toluene complex (2:1) 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Copper(I) trifluoromethanesulphonate hemitoluene complex
L19656.03 L19656.06
L19656.03EA 1550 SEK
L19656.03 L19656.06
Copper(I) trifluoromethanesulphonate toluene complex (2:1) 98%
Copper(I) trifluoromethanesulphonate toluene complex
Purity: 98% CAS No.: 48209-28-5 Molecular Formula: [CF3SO3Cu]2.C6H5CH3 Formula Weight: 517.37 Flash Point: 76°(168°F) Melting Point: 180° dec. UN No.: UN1325 MDL No.: MFCD01863766
Formel: [CF₃SO₃Cu]₂·C₆H₅CH₃
MW: 517,37 g/mol
Smältpunkt: 180 °C (dec.)
Flampunkt: 76 °C (168 °F)
Storage Temperature: Omgivande
MDL Number: MFCD01863766
CAS nummer: 48209-28-5
UN: 1325
ADR: 4.1,III