N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 94%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Purity: 94%
CAS No.: 93102-05-7
Molecular Formula: C13H23NOSi
Formula Weight: 237.42
Storage Temperature: 2°C to 8°C
Flash Point: 66°(150°F)
Refractive Index: 1.4920
Boiling Point: 76°/0.3mm
Beilstein Reference: 4311216
MDL No.: MFCD00674005
Formel:
C₁₃H₂₃NOSi Kokpunkt: 76 °C (0,3 mmHg) Densitet: 0,928 Flampunkt: 66 °C (150 °F) |
MDL Number:
MFCD00674005 CAS nummer: 93102-05-7 |