4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Methoxy-3-pyrrolin-2-one, 4-Methoxy-1H-pyrrol-2(5H)-one
L19344.03 L19344.06
L19344.03EA 1775 SEK
L19344.03 L19344.06
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 99%
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one
Purity: 99% CAS No.: 69778-83-2 Molecular Formula: C5H7NO2 Formula Weight: 113.12 Melting Point: 130-134° Beilstein Reference: 1524505 MDL No.: MFCD00071564
Formel: C₅H₇NO₂
MW: 113,12 g/mol
Smältpunkt: 130…134 °C
Storage Temperature: Omgivande
MDL Number: MFCD00071564
CAS nummer: 69778-83-2