(1R)-(-)-Menthyl glyoxylate monohydrate 98%
Leverantör: Thermo Scientific
Fara
L19208.14 L19208.22
L19208.14EA 1875 SEK
L19208.14 L19208.22
(1R)-(-)-Menthyl glyoxylate monohydrate 98%
(1R)-(-)-Menthyl glyoxylate monohydrate
Purity: 98% CAS No.: 111969-64-3 Molecular Formula: C12H22O4 Formula Weight: 230.31 (212.29anhy) Melting Point: 76-80° Boiling Point: 95-96°/0.3mm Beilstein Reference: 5637371 Optical Rotation: -75° (c=1 in methanol) UN No.: UN3077 MDL No.: MFCD00792503
Formel: C₁₂H₂₂O₄
MW: 230,31 g/mol (212,29 anhy)
Kokpunkt: 95…96 °C (0,3 mmHg)
Smältpunkt: 76…80 °C
Densitet: 1,182
Storage Temperature: Omgivande
MDL Number: MFCD00792503
CAS nummer: 111969-64-3
EINECS: 416-810-6
ADR: 9,III