1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: [1,2,4]Triazolo[4,3-a]pyridin-3(2H)-one, 3-Hydroxy-1,2,4-triazolo[4,3-a]pyridine
L18705.06 L18705.14
L18705.06EA 660 SEK
L18705.06 L18705.14
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥98%
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one
Purity: 98+% CAS No.: 6969-71-7 Molecular Formula: C6H5N3O Formula Weight: 135.13 Melting Point: 233-235° Beilstein Reference: 607433 MDL No.: MFCD00022632
Formel: C₆H₅N₃O
MW: 135,13 g/mol
Smältpunkt: 233…235 °C
Storage Temperature: Omgivande
MDL Number: MFCD00022632
CAS nummer: 6969-71-7
EINECS: 230-191-8