O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: N-(6-cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium tetrafluoroborate, TOTU
L18496.06 L18496.14
L18496.06EA 1825 SEK
L18496.06 L18496.14
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate
Purity: 97% CAS No.: 136849-72-4 Molecular Formula: C10H17BF4N4O3 Formula Weight: 328.08 Melting Point: 143-146° dec. MDL No.: MFCD00192127
Formel: C₁₀H₁₇BF₄N₄O₃
MW: 328,07 g/mol
Smältpunkt: 143…146 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00192127
CAS nummer: 136849-72-4