Bis[2-(diphenylphosphino)phenyl]ether 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2'-Bis(diphenylphosphino)diphenyl Ether, DPEphos, Bis(2-diphenylphosphinophenyl)ether, (Oxydi-2,1-phenylene)bis(diphenylphosphine), (Oxybis(2,1-phenylene))bis(diphenylphosphine)
L18481.03 L18481.06
L18481.03EA 750 SEK
L18481.03 L18481.06
Bis[2-(diphenylphosphino)phenyl]ether 98%
Bis[2-(diphenylphosphino)phenyl]ether
Purity: 98% CAS No.: 166330-10-5 Molecular Formula: C36H28OP2 Formula Weight: 538.57 Melting Point: 184-186° Beilstein Reference: 7729718 MDL No.: MFCD00233863
Formel: C₃₆H₂₈OP₂
MW: 538,57 g/mol
Smältpunkt: 184…186 °C
Storage Temperature: Omgivande
MDL Number: MFCD00233863
CAS nummer: 166330-10-5