1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone ≥98%
Leverantör: Thermo Scientific
Synonyms: (3aR,3bS,6S,6aR,7aR)-6-Hydroxy-2,2-dimethyltetrahydrofuro[2',3':4,5]furo[2,3-d][1,3]dioxol-5(3aH)-one
L17765.09 L17765.18
L17765.09EA 1375 SEK
L17765.09 L17765.18
1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone ≥98%
1,2-O-Isopropylidene-alpha-D-glucurono-6,3-lactone
Purity: 98+% CAS No.: 20513-98-8 Molecular Formula: C9H12O6 Formula Weight: 216.19 Melting Point: 119-121° Optical Rotation: +52° (c=1 in chloroform) MDL No.: MFCD00061641
Formel: C₉H₁₂O₆
MW: 216,19 g/mol
Smältpunkt: 119…121 °C
Storage Temperature: Omgivande
MDL Number: MFCD00061641
CAS nummer: 20513-98-8