tert-Butyl-1,4-diazepane-1-carboxylate 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-BOC-hexahydro-1,4-diazepine, tert-Butyl 1,4-diazepane-1-carboxylate
L17728.03 L17728.06
L17728.03EA 1050 SEK
L17728.03 L17728.06
tert-Butyl-1,4-diazepane-1-carboxylate 98%
tert-Butyl-1,4-diazepane-1-carboxylate
Purity: 98% CAS No.: 112275-50-0 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Flash Point: 110°(230°F) Refractive Index: 1.4710 Boiling Point: 90°/0.5mm Beilstein Reference: 7581789 UN No.: UN2735 MDL No.: MFCD00276987
Formel: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Kokpunkt: 90 °C (0,5 mmHg)
Densitet: 1,016
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00276987
CAS nummer: 112275-50-0
ADR: 8,III