Dimethyl-2,6-naphthalenedicarboxylate ≥99%
Leverantör: Thermo Scientific
Synonyms: Dimethyl 2,6-naphthalenedicarboxylate, 2,6-Naphthalenedicarboxylic Acid Dimethyl Ester
L17689.22 L17689.06 L17689.14
L17689.22EA 25500 SEK
L17689.22 L17689.06 L17689.14
Dimethyl-2,6-naphthalenedicarboxylate ≥99%
Dimethyl-2,6-naphthalenedicarboxylate
Purity: 99+% CAS No.: 840-65-3 Molecular Formula: C14H12O4 Formula Weight: 244.25 Flash Point: 232°(449°F) Melting Point: 187-193° Beilstein Reference: 987259 MDL No.: MFCD00004100
Formel: C₁₄H₁₂O₄
MW: 244,25 g/mol
Smältpunkt: 187…193 °C
Flampunkt: 232 °C (449 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004100
CAS nummer: 840-65-3
EINECS: 212-661-4