2-Hydroxy-2-(trifluoromethyl)propionic acid 94%
Leverantör: Thermo Fisher Scientific
Fara
L17399.03 L17399.06
L17399.03EA 800 SEK
L17399.03 L17399.06
2-Hydroxy-2-(trifluoromethyl)propionic acid 94%
2-Hydroxy-2-(trifluoromethyl)propionic acid
Purity: 94% CAS No.: 114715-77-4 Molecular Formula: C4H5F3O3 Formula Weight: 158.08 Melting Point: ca 85° Boiling Point: 189° Beilstein Reference: 1723691 UN No.: UN3261 MDL No.: MFCD00190646
Formel: C₄H₅F₃O₃
MW: 158.08 g/mol
Kokpunkt: 189 °C
Smältpunkt: ∼ 85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00190646
CAS nummer: 114715-77-4
ADR: 8 II