3-(Trifluoromethyl)-3-hydroxybutyric acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 4,4,4-Trifluoro-3-hydroxy-3-methylbutanoic acid, 3-Hydroxy-3-(trifluoromethyl)butyric acid
L16873.03 L16873.06
L16873.03EA 1425 SEK
L16873.03 L16873.06
3-(Trifluoromethyl)-3-hydroxybutyric acid 98%
3-(Trifluoromethyl)-3-hydroxybutyric acid
Purity: 98% CAS No.: 338-03-4 Molecular Formula: C5H7F3O3 Formula Weight: 172.10 Refractive Index: 1.3880 Melting Point: 39-41° Boiling Point: 87-90°/0.5mm UN No.: UN3261 MDL No.: MFCD00039531
Formel: C₅H₇F₃O₃
MW: 172,1 g/mol
Kokpunkt: 87…90 °C (0,5 mmHg)
Smältpunkt: 39…41 °C
Storage Temperature: Omgivande
MDL Number: MFCD00039531
CAS nummer: 338-03-4
ADR: 8,III