5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 5-Nitro-1H-benzo[d]imidazol-2(3H)-one
L16391.06 L16391.14
L16391.06EA 1950 SEK
L16391.06 L16391.14
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 99%
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one
Purity: 99% CAS No.: 93-84-5 Molecular Formula: C7H5N3O3 Formula Weight: 179.14 Melting Point: 300° Beilstein Reference: 171386 UN No.: UN2811 MDL No.: MFCD00220274
Formel: C₇H₅N₃O₃
MW: 179,14 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00220274
CAS nummer: 93-84-5
EINECS: 202-282-2
ADR: 6.1,III