2-Isocyanato-1-methoxy-4-methylbenzene 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Methoxy-5-methylphenyl isocyanate, 2-Methoxy-5-methylphenylisocyanate
L16001.03 L16001.06
L16001.03EA 510 SEK
L16001.03 L16001.06
2-Isocyanato-1-methoxy-4-methylbenzene 97%
2-Isocyanato-1-methoxy-4-methylbenzene
Purity: 97% CAS No.: 59741-04-7 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Flash Point: 110°(230°F) Refractive Index: 1.5370 Boiling Point: 240° Beilstein Reference: 3604599 UN No.: UN2206 MDL No.: MFCD00013869
Formel: C₉H₉NO₂
MW: 163,18 g/mol
Kokpunkt: 240 °C
Densitet: 1,112
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013869
CAS nummer: 59741-04-7
ADR: 6.1,III