1-(2-Pyrimidyl)piperazine 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-(1-Piperazinyl)pyrimidine, 2-(Piperazin-1-yl)pyrimidine
L15884.06 L15884.14 L15884.22
L15884.06EA 710 SEK
L15884.06 L15884.14 L15884.22
1-(2-Pyrimidyl)piperazine 99%
1-(2-Pyrimidyl)piperazine
Purity: 99% CAS No.: 20980-22-7 Molecular Formula: C8H12N4 Formula Weight: 164.21 Melting Point: 31-35° Boiling Point: 118-120°/2mm Beilstein Reference: 151178 UN No.: UN3259 MDL No.: MFCD00040742
Formel: C₈H₁₂N₄
Kokpunkt: 118…120 °C (2 mmHg)
Smältpunkt: 31…35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040742
CAS nummer: 20980-22-7
EINECS: 244-135-5
ADR: 8,III