4-Chloro-3-nitro-2-pyridone 98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
L15686.14
L15686.06
L15686.14EA
8850
SEK
L15686.14
L15686.06
4-Chloro-3-nitro-2-pyridone 98%
4-Chloro-3-nitro-2-pyridone
Purity: 98%
CAS No.: 165547-79-5
Molecular Formula: C5H3ClN2O3
Formula Weight: 174.54
Melting Point: 222-224°
Beilstein Reference: 149557
MDL No.: MFCD03093067
Formel:
C₅H₃ClN₂O₃ Smältpunkt: 222…224 °C Storage Temperature: Omgivande |
MDL Number:
MFCD03093067 CAS nummer: 165547-79-5 |