Phosphorus pentoxide - methanesulphonic acid (∼7,5% P₂O₅) w/w
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Eaton's Reagent
L15607.AE L15607.AP
L15607.AEEA 1025 SEK
L15607.AE L15607.AP
Phosphorus pentoxide - methanesulphonic acid (∼7,5% P₂O₅) w/w
Phosphorus pentoxide - methanesulphonic acid
CAS No.: 39394-84-8 Molecular Formula: O5P2 Formula Weight: 141.95 Flash Point: 110°(230°F) Refractive Index: 1.4350 UN No.: UN3265 MDL No.: MFCD00070545
MW: 141.95 g/mol
Densitet: 1.515
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00070545
CAS nummer: 39394-84-8
UN: 3265
ADR: 8 II