4'-Chloro-2,2':6',2''-terpyridine 98%
Leverantör: Thermo Scientific
|
Varning
|
L14728.03
L14728.MD
L14728.03EA
3925
SEK
L14728.03
L14728.MD
4'-Chloro-2,2':6',2''-terpyridine 98%
4'-Chloro-2,2':6',2''-terpyridine
Purity: 98%
CAS No.: 128143-89-5
Molecular Formula: C15H10ClN3
Formula Weight: 267.72
Melting Point: 148-152°
Beilstein Reference: 3613386
MDL No.: MFCD00191930
Formel:
C₁₅H₁₀ClN₃ MW: 267,716 g/mol Smältpunkt: 148…152 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00191930 CAS nummer: 128143-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...