(R)-(-)-Tetrahydrofurfurylamine ≥98%
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
(R)-(-)-2-(Aminomethyl)tetrahydrofuran, (R)-2-Tetrahydrofurfurylamine
L14410.03
L14410.MD
L14410.03EA
2350
SEK
L14410.03
L14410.MD
(R)-(-)-Tetrahydrofurfurylamine ≥98%
(R)-(-)-Tetrahydrofurfurylamine
Purity: 98+%
CAS No.: 7202-43-9
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 45°(113°F)
Refractive Index: 1.4550
Boiling Point: 152-153°
Beilstein Reference: 4652873
Optical Rotation: -12° (c=2 in chloroform)
UN No.: UN2943
MDL No.: MFCD00192476
Formel:
C₅H₁₁NO MW: 101.15 g/mol Kokpunkt: 152…153 °C Densitet: 0.980 Flampunkt: 45 °C (113 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00192476 CAS nummer: 7202-43-9 UN: 2943 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...