Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,1,1-Triphenyl-N-(triphenylphosphoranylidene)phosphoraniminium chloride, Bis(triphenylphosphine)iminium chloride
L14339.06 L14339.14
L14339.06EA 1850 SEK
L14339.06 L14339.14
Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97%
Bis(triphenylphosphoranylidene)ammonium chloride
Purity: 97% CAS No.: 21050-13-5 Molecular Formula: C36H30ClNP2 Formula Weight: 574.05 Melting Point: 274-276° Beilstein Reference: 4097979 MDL No.: MFCD00151523
Formel: C₃₆H₃₀ClNP₂
MW: 574.041 g/mol
Smältpunkt: 274…276 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151523
CAS nummer: 21050-13-5
EINECS: 244-170-6