Dowex® 1X8 (Cl) 200-400 mesh
Leverantör: Thermo Fisher Scientific
L14258.18 L14258.30 L14258.0B
L14258.18EA 1275 SEK
L14258.18 L14258.30 L14258.0B
Dowex® 1X8 (Cl) 200-400 mesh
Dowex® 1X8 (Cl)
CAS No.: 12627-85-9 Odour: Amine-like MDL No.: MFCD00132718
Storage Temperature: Omgivande
MDL Number: MFCD00132718
CAS nummer: 12627-85-9