Dowex® 1X8 (Cl) 100-200 mesh
Leverantör: Thermo Fisher Scientific
L14257.18
L14257.30
L14257.0B
L14257.18EA
1325
SEK
L14257.18
L14257.30
L14257.0B
Dowex® 1X8 (Cl) 100-200 mesh
Dowex® 1X8 (Cl)
CAS No.: 12627-85-9
Odour: Amine-like
MDL No.: MFCD00132718
Storage Temperature:
Omgivande |
MDL Number:
MFCD00132718 CAS nummer: 12627-85-9 |