(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 99%
Leverantör: Thermo Scientific
Synonyms: (1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol
L14053.03 L14053.06 L14053.14
L14053.03EA 720 SEK
L14053.03 L14053.06 L14053.14
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 99%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
Purity: 99% CAS No.: 18680-27-8 Molecular Formula: C10H18O2 Formula Weight: 170.25 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 56-58° Beilstein Reference: 1853591 Optical Rotation: +8.7° (c=6.5 in toluene) MDL No.: MFCD00077851
Formel: C₁₀H₁₈O₂
MW: 170,25 g/mol
Smältpunkt: 56…58 °C
Flampunkt: >110 °C (230 °F)
MDL Number: MFCD00077851
CAS nummer: 18680-27-8