Dowex® 50WX2 (H) 200-400 mesh
Leverantör: Thermo Fisher Scientific
L13945.18 L13945.30 L13945.0B
L13945.18EA 1250 SEK
L13945.18 L13945.30 L13945.0B
Dowex® 50WX2 (H) 200-400 mesh
Dowex® 50WX2 (H)
CAS No.: 12612-37-2 MDL No.: MFCD00132722
Storage Temperature: Omgivande
MDL Number: MFCD00132722
CAS nummer: 12612-37-2