Dowex® 50WX8 (H) 100-200 mesh
Leverantör: Thermo Fisher Scientific
Varning
L13921.18 L13921.30 L13921.0B
L13921.18EA 1500 SEK
L13921.18 L13921.30 L13921.0B
Dowex® 50WX8 (H) 100-200 mesh
Dowex® 50WX8 (H)
CAS No.: 11119-67-8 MDL No.: MFCD00132726
Storage Temperature: Omgivande
MDL Number: MFCD00132726
CAS nummer: 11119-67-8