2-Amino-5-n-propylsulphonylbenzimidazole ≥98%

Leverantör: Thermo Fisher Scientific

L13711.MC L13711.03
L13711.MCEA 1150 SEK
L13711.MC L13711.03
2-Amino-5-n-propylsulphonylbenzimidazole ≥98%
2-Amino-5-n-propylsulphonylbenzimidazole

Purity: 98+% CAS No.: 80983-34-2 Molecular Formula: C10H13N3O2S Formula Weight: 239.30 Melting Point: 220-222° MDL No.: MFCD01075656

Formel: C₁₀H₁₃N₃O₂S
Smältpunkt: 220…222 °C
Storage Temperature: Omgivande
MDL Number: MFCD01075656
CAS nummer: 80983-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR