3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: [3,5-Bis(trifluoromethyl)phenyl]chloro(dimethyl)silane
L13709.03 L13709.06
L13709.03EA 1650 SEK
L13709.03 L13709.06
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane 95%
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
Purity: 95% CAS No.: 732306-23-9 Molecular Formula: C10H9ClF6Si Formula Weight: 306.71 Refractive Index: 1.4240 Boiling Point: 95-96°/30mm UN No.: UN3265 MDL No.: MFCD01862186
Formel: C₁₀H₉ClF₆Si
MW: 306.71 g/mol
Kokpunkt: 95…96 °C (30 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD01862186
CAS nummer: 732306-23-9
UN: 3265
ADR: 8 III