Methacroylcholine chloride ≥72% i vattenlösning stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(Methacryloyloxy)-N,N,N-trimethylethanaminium chloride, (Methacryloyloxyethyl)trimethyl ammonium chloride, 2-(Methacryloyloxy)ethyltrimethylammonium chloride, Trimethyl-2-methacroyloxyethylammonium Chloride
Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 150-200 ppm
L13523.22 L13523.36
L13523.22EA 685 SEK
L13523.22 L13523.36
Methacroylcholine chloride ≥72% i vattenlösning stabilised
Methacroylcholine chloride
CAS No.: 5039-78-1 Molecular Formula: C9H18ClNO2 Formula Weight: 207.70 Refractive Index: 1.4660 Melting Point: -25° Boiling Point: 100° Beilstein Reference: 8168395 MDL No.: MFCD00060097
Formel: C₉H₁₈ClNO₂
MW: 207,70 g/mol
Kokpunkt: >100 °C
Smältpunkt: –25 °C
Densitet: 1,105
Storage Temperature: Omgivande
MDL Number: MFCD00060097
CAS nummer: 5039-78-1
EINECS: 225-733-5