cis-1,4-Diacetoxy-2-butene 96%
Leverantör: Thermo Fisher Scientific
Synonyms: cis-2-Butene-1,4-diol Diacetate, (Z)-But-2-ene-1,4-diyl diacetate
L13024.09 L13024.18
L13024.09EA 1100 SEK
L13024.09 L13024.18
cis-1,4-Diacetoxy-2-butene 96%
cis-1,4-Diacetoxy-2-butene
Purity: 96% CAS No.: 25260-60-0 Molecular Formula: C8H12O4 Formula Weight: 172.18 Flash Point: 110°(230°F) Refractive Index: 1.4416 Boiling Point: 120-121°/18mm Beilstein Reference: 1724379 MDL No.: MFCD00059339
Formel: C₈H₁₂O₄
Kokpunkt: 120…121 °C (18 mmHg)
Densitet: 1,090
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00059339
CAS nummer: 25260-60-0