1-Benzyl-5-phenylbarbituric acid ≥98%
Leverantör: Thermo Scientific
L12954.06
L12954.14
L12954.06EA
545
SEK
L12954.06
L12954.14
1-Benzyl-5-phenylbarbituric acid ≥98%
1-Benzyl-5-phenylbarbituric acid
Purity: 98+%
CAS No.: 72846-00-5
Molecular Formula: C17H14N2O3
Formula Weight: 294.31
Melting Point: 163-165°
MDL No.: MFCD00082750
Formel:
C₁₇H₁₄N₂O₃ Smältpunkt: 163…165 °C |
MDL Number:
MFCD00082750 CAS nummer: 72846-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...