Diacetin mixture of isomers ∼50% remainder triacetin and monoacetin, tech.
Leverantör: Thermo Scientific
Synonyms: Glycerol alpha,alpha'-Diacetate, 2-Hydroxypropane-1,3-diyl diacetate
L12708.30 L12708.0B
L12708.30EA 885 SEK
L12708.30 L12708.0B
Diacetin mixture of isomers ∼50% remainder triacetin and monoacetin, tech.
Diacetin
CAS No.: 25395-31-7 Molecular Formula: C7H12O5 Formula Weight: 176.17 Flash Point: 141°(285°F) Refractive Index: 1.4440 Boiling Point: 250-280° dec. Beilstein Reference: 1706903 MDL No.: MFCD00008717
Formel: C₇H₁₂O₅
MW: 176,17 g/mol
Kokpunkt: 250…280 °C (dec.)
Densitet: 1,190
Flampunkt: 141 °C (285 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008717
CAS nummer: 25395-31-7
EINECS: 246-941-2
Merck Index: 14,2961