TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥97%
Leverantör: Thermo Scientific
Synonyms: Tetrabromophenolphthalein Ethyl Ester Potassium Salt
L12674.03 L12674.06
L12674.03EA 2325 SEK
L12674.03 L12674.06
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥97%
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt)
CAS No.: 62637-91-6 Molecular Formula: C22H13Br4KO4 Formula Weight: 700.06 Melting Point: ca 273° dec. Beilstein Reference: 3848263 MDL No.: MFCD00011662
Formel: C₂₂H₁₄Br₄O₄
MW: 700,06 g/mol
Smältpunkt: ∼ 273 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00011662
CAS nummer: 62637-91-6
EINECS: 263-661-6