2-Bromoethyl acrylate 94% stabilised
Leverantör: Thermo Scientific
|
Varning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 900 - 1500 ppm
L12502.06
L12502.03
L12502.06EA
3400
SEK
L12502.06
L12502.03
2-Bromoethyl acrylate 94% stabilised
2-Bromoethyl acrylate
Stab. with 0.1% 4-methoxyphenol
Purity: 94%
CAS No.: 4823-47-6
Molecular Formula: CH2=CHCO2CH2CH2Br
Formula Weight: 179.02
Storage Temperature: 2°C to 8°C
Boiling Point: 52-53°/5mm
Beilstein Reference: 1749713
MDL No.: MFCD00013541
Formel:
CH₂=CHCO₂CH₂CH₂Br MW: 179,02 g/mol Kokpunkt: 52…53 °C (5 mmHg) |
MDL Number:
MFCD00013541 CAS nummer: 4823-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...