1-(4-Fluorophenyl)-2-methyl-2-propylamine 96%
Leverantör: Thermo Scientific
Fara
L11858.03 L11858.06
L11858.03EA 2025 SEK
L11858.03 L11858.06
1-(4-Fluorophenyl)-2-methyl-2-propylamine 96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine
Purity: 96% CAS No.: 1200-27-7 Molecular Formula: C10H14FN Formula Weight: 167.23 Flash Point: 110°(230°F) Refractive Index: 1.4950 Boiling Point: 82-83°/9mm Beilstein Reference: 2802368 UN No.: UN2735 MDL No.: MFCD00082702
Formel: C₁₀H₁₄FN
MW: 167,227 g/mol
Kokpunkt: 82…83 °C (9 mmHg)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00082702
CAS nummer: 1200-27-7
ADR: 8,III