Hexyl methacrylate 97% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Methacrylic Acid Hexyl Ester
Stabilization: 100 ppm Stabilised with 4-methoxyphenol (MEHQ)
L11569.14 L11569.22
L11569.14EA 1050 SEK
L11569.14 L11569.22
Hexyl methacrylate 97% stabilised
Hexyl methacrylate
Stab. with 100ppm 4-methoxyphenol
Purity: 97% CAS No.: 142-09-6 Molecular Formula: C10H18O2 Formula Weight: 170.25 Flash Point: 80°(176°F) Refractive Index: 1.4310 Odour: Characteristic Melting Point: -57° Boiling Point: 204° Beilstein Reference: 1754703 MDL No.: MFCD00015283
Formel: C₁₀H₁₈O₂
MW: 170.25 g/mol
Kokpunkt: 204 °C
Smältpunkt: –57 °C
Densitet: 0.885
Flampunkt: 80 °C (176 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00015283
CAS nummer: 142-09-6
EINECS: 205-521-9