4-Hydroxy-6-methyl-2-pyrone 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Hydroxy-6-methyl-2H-pyran-2-one, 3,5-Dihydroxysorbic acid d-lactone, 2-hydroxy-6-methyl-pyran-4-one
L11457.14 L11457.22
L11457.14EA 1450 SEK
L11457.14 L11457.22
4-Hydroxy-6-methyl-2-pyrone 98%
4-Hydroxy-6-methyl-2-pyrone
Purity: 98% CAS No.: 675-10-5 Molecular Formula: C6H6O3 Formula Weight: 126.11 Melting Point: ca 186° dec. Beilstein Reference: 113815 MDL No.: MFCD00006641
Formel: C₆H₆O₃
MW: 126,11 g/mol
Smältpunkt: ∼ 186 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006641
CAS nummer: 675-10-5
EINECS: 211-619-2