Dimethyl-cis-stilbene-4,4'-dicarboxylate
Leverantör: Thermo Scientific
L11142.03 L11142.MD
L11142.03EA 2000 SEK
L11142.03 L11142.MD
Dimethyl-cis-stilbene-4,4'-dicarboxylate
Dimethyl-cis-stilbene-4,4'-dicarboxylate
CAS No.: 143130-82-9 Molecular Formula: C18H16O4 Formula Weight: 296.33 Melting Point: 112-114° Beilstein Reference: 2386934 MDL No.: MFCD00082669
Formel: C₁₈H₁₆O₄
MW: 296,33 g/mol
Smältpunkt: 112…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082669
CAS nummer: 143130-82-9