Vinyl chloroacetate 99% stabilised
Leverantör: Thermo Fisher Scientific
Fara
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
L09446.09 L09446.18
L09446.09EA 1075 SEK
L09446.09 L09446.18
Vinyl chloroacetate 99% stabilised
Vinyl chloroacetate
Stab. with 4-methoxyphenol
Purity: 99% CAS No.: 2549-51-1 Molecular Formula: ClCH2CO2CH=CH2 Formula Weight: 120.54 Flash Point: 51°(123°F) Refractive Index: 1.4435 Odour: Pungent Boiling Point: 136° Beilstein Reference: 1741679 UN No.: UN2589 MDL No.: MFCD00045275
Formel: C₄H₅ClO₂
Kokpunkt: 136 °C
Densitet: 1,190
Flampunkt: 51 °C (123 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00045275
CAS nummer: 2549-51-1
EINECS: 219-834-3
UN: 2589
ADR: 6.1,II