5,5-Dimethyl-1,3-dioxan-2-one, tech.
Leverantör: Thermo Scientific
L08767.06 L08767.03
L08767.06EA 3250 SEK
L08767.06 L08767.03
5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one
CAS No.: 3592-12-9 Molecular Formula: C6H10O3 Formula Weight: 130.14 Melting Point: 105-114° Beilstein Reference: 109055 MDL No.: MFCD00014650
Formel: C₆H₁₀O₃
MW: 130,14 g/mol
Smältpunkt: 105…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014650
CAS nummer: 3592-12-9