3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride
L08750.14 L08750.22
L08750.14EA 890 SEK
L08750.14 L08750.22
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Purity: 98% CAS No.: 4568-71-2 Molecular Formula: C13H16ClNOS Formula Weight: 269.80 Melting Point: 142-145° Beilstein Reference: 3788921 MDL No.: MFCD00011959
Formel: C₁₃H₁₆ClNOS
MW: 269,79 g/mol
Smältpunkt: 142…145 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011959
CAS nummer: 4568-71-2
EINECS: 224-947-6