N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
Leverantör: Thermo Fisher Scientific
L08498.03
L08498.06
L08498.03EA
570
SEK
L08498.03
L08498.06
N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
N-(tert-Butoxycarbonyl)-L-aspartic acid
Purity: 98%
CAS No.: 13726-67-5
Molecular Formula: C9H15NO6
Formula Weight: 233.23
Storage Temperature: 2°C to 8°C
Melting Point: 115°
Beilstein Reference: 1913973
Optical Rotation: -5.5° (c=1 in methanol)
MDL No.: MFCD00037279
Formel:
C₉H₁₅NO₆ Smältpunkt: 115 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00037279 CAS nummer: 13726-67-5 EINECS: 237-294-7 |