Norbornane-2-carboxylic acid predominantly endo isomer 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Bicyclo[2.2.1]heptane-2-carboxylic acid
L08439.06 L08439.14
L08439.06EA 1900 SEK
L08439.06 L08439.14
Norbornane-2-carboxylic acid predominantly endo isomer 98%
Norbornane-2-carboxylic acid
Purity: 98% CAS No.: 824-62-4 Molecular Formula: C8H12O2 Formula Weight: 140.18 Flash Point: 100°(212°F) Refractive Index: 1.4855 Melting Point: ca 40° Boiling Point: 134-136°/16mm Beilstein Reference: 2042631 UN No.: UN3261 MDL No.: MFCD00167749
Formel: C₈H₁₂O₂
MW: 140,18 g/mol
Kokpunkt: 134…136 °C (16 mmHg)
Smältpunkt: ∼ 40 °C
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00167749
CAS nummer: 824-62-4
EINECS: 212-532-2
UN: 3261
ADR: 8,III