1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 97%

Leverantör: Thermo Scientific
Varning

Synonyms: 3-(3,5-Dimethyl-1H-pyrazol-1-yl)-3-oxopropanenitrile

L08369.06 L08369.14
L08369.06EA 2100 SEK
L08369.06 L08369.14
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 97%
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole

Purity: 97% CAS No.: 36140-83-7 Molecular Formula: C8H9N3O Formula Weight: 163.18 Melting Point: 118-122° Beilstein Reference: 6286 MDL No.: MFCD00051954

Formel: C₈H₉N₃O
MW: 163,18 g/mol
Smältpunkt: 118…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051954
CAS nummer: 36140-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR