Lithium aluminium di-n-butylamide lösning 0.16 M i 1,2-Dimetoxietan
Leverantör: Thermo Scientific
Varning
Synonyms: Lithium tetrakis(N-butyl-1-butanaminato)aluminate(1-), Aluminate(1-), tetrakis(N-butyl-1-butanaminato)-, lithium (1:1)
L08269.AE L08269.AP
L08269.AEEA 3200 SEK
L08269.AE L08269.AP
Lithium aluminium di-n-butylamide lösning 0.16 M i 1,2-Dimetoxietan
Lithium aluminium di-n-butylamide
CAS No.: 15405-86-4 Molecular Formula: C32H72AlLiN4 Formula Weight: 546.87 Storage Temperature: 2°C to 8°C Melting Point: -54° UN No.: UN1993 MDL No.: MFCD00051918
Formel: C₃₂H₇₂AlLiN₄
MW: 546,87 g/mol
Smältpunkt: –54 °C
Densitet: 0,860
Storage Temperature: Kylskåp
MDL Number: MFCD00051918
CAS nummer: 15405-86-4
UN: 1993
ADR: 3,III