Acetazolamide 99%
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
2-Acetamido-5-sulphonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulphonamide
L07562.06
L07562.14
L07562.06EA
815
SEK
L07562.06
L07562.14
Acetazolamide 99%
Acetazolamide
Purity: 99%
CAS No.: 59-66-5
Molecular Formula: C4H6N4O3S2
Formula Weight: 222.25
Melting Point: ca 255° dec.
Beilstein Reference: 212994
MDL No.: MFCD00003105
Formel:
C₄H₆N₄O₃S₂ MW: 222.24 g/mol Smältpunkt: ∼ 255 °C (dec.) Storage Temperature: Omgivande |
MDL Number:
MFCD00003105 CAS nummer: 59-66-5 EINECS: 200-440-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...