2-(2-Aminophenyl)benzimidazole 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-(1H-Benzimidazol-2-yl)aniline
L07034.06 L07034.14
L07034.06EA 1025 SEK
L07034.06 L07034.14
2-(2-Aminophenyl)benzimidazole 98%
2-(2-Aminophenyl)benzimidazole
Purity: 98% CAS No.: 5805-39-0 Molecular Formula: C13H11N3 Formula Weight: 209.24 Melting Point: 212-216° Beilstein Reference: 173018 MDL No.: MFCD00051496
Formel: C₁₃H₁₁N₃
MW: 209,252 g/mol
Smältpunkt: 212…216 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051496
CAS nummer: 5805-39-0
EINECS: 227-361-9