3,5-Pyrazoledicarboxylic acid monohydrate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1H-Pyrazole-3,5-dicarboxylic acid hydrate, Pyrazole-3,5-dicarboxylic acid monohydrate, 1H-Pyrazole-3,5-dicarboxylic acid monohydrate
L06731.06 L06731.14 L06731.22
L06731.06EA 1550 SEK
L06731.06 L06731.14 L06731.22
3,5-Pyrazoledicarboxylic acid monohydrate 98%
3,5-Pyrazoledicarboxylic acid monohydrate
Purity: 98% CAS No.: 303180-11-2 Molecular Formula: C5H4N2O4.H2O Formula Weight: 174.11 (156.09anhy) Melting Point: ca 294° dec. Beilstein Reference: 147821 MDL No.: MFCD00149323
Formel: C₅H₄N₂O₄·H₂O
MW: 174,11 g/mol
Smältpunkt: ∼ 294 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00149323
CAS nummer: 303180-11-2
EINECS: 221-474-7